Learn More
Electron Microscopy Sciences Platinum Line Coverglass Rectangle #1, 24 x 60mm 1 OZ
SDP
Catalog No. 501928969
Encompass_Preferred
Platinum Line No. 1 Coverglass are made of a water-white colorless borosilicate glass, with an optimum hydrolytic resistance. Ultra-thin. Performs exceptionally well on all automated glass cverslippers.
Thickness: #1
Size: 24 x 60 mm
Catalog No. 50-192-8969 Supplier Electron Microscopy Sciences Supplier No. 7188723
Platinum Line No. 1 Coverglass are made of a water-white colorless borosilicate glass, with an optimum hydrolytic resistance. Ultra-thin. Performs exceptionally well on all automated glass cverslippers.
Thickness: #1
Size: 24 x 60 mm