Learn More
Electron Microscopy Sciences Tanner Coverglass Rectangle #1, 24 x 55 mm 10 OZ
SDP
Catalog No. 501928946
Encompass_Preferred
Tanner coverglass is suitable for a host of laboratory applications ranging from histopathology, cytology, microbiology and life sciences. It is ultrathin and disposable
Thickness: #1
Size: 24 x 55 mm
Catalog No. 50-192-8946 Supplier Electron Microscopy Sciences Supplier No. 7188464
Tanner coverglass is suitable for a host of laboratory applications ranging from histopathology, cytology, microbiology and life sciences. It is ultrathin and disposable
Thickness: #1
Size: 24 x 55 mm