Learn More
Medchemexpress LLC CVT-10216 | 1005334-57-5 | 99.9% | 465.48 | 50 MG
SDP
Catalog No. 5000402802
Encompass_Preferred
CVT-10216 is a highly selective, reversible aldehyde dehydrogenase-2 (ALDH-2) inhibitor with an IC50 of 29 nM. CVT-10216 also has inhibitory effect of ALDH-1 with an IC50 of 1.3 μM. CVT-10216 can reduce excessive alcohol drinking in alcohol-preferring rats and exhibit anxiolytic effects.
  • Highly selective, reversible aldehyde dehydrogenase-2 (ALDH-2) inhibitor with an IC50 of 29 nM.
  • Inhibitory effect on ALDH-1 with an IC50 of 1.3 μM.
  • Reduces excessive alcohol drinking in alcohol-preferring rats.
  • Exhibits anxiolytic effects.
Catalog No. 50-004-02802 Supplier Medchemexpress LLC Supplier No. HY1980150MG
CVT-10216 is a highly selective, reversible aldehyde dehydrogenase-2 (ALDH-2) inhibitor with an IC50 of 29 nM. CVT-10216 also has inhibitory effect of ALDH-1 with an IC50 of 1.3 μM. CVT-10216 can reduce excessive alcohol drinking in alcohol-preferring rats and exhibit anxiolytic effects.
  • Highly selective, reversible aldehyde dehydrogenase-2 (ALDH-2) inhibitor with an IC50 of 29 nM.
  • Inhibitory effect on ALDH-1 with an IC50 of 1.3 μM.
  • Reduces excessive alcohol drinking in alcohol-preferring rats.
  • Exhibits anxiolytic effects.