Learn More
Medchemexpress LLC HY-19801 5mg Medchemexpress, CVT-10216 CAS:1005334-57-5 Purity:>98%
SDP
Catalog No. 501970037
Encompass_Preferred
Medchemexpress, HY-19801 5mg CVT-10216 CAS:1005334-57-5 CVT-10216 is a highly selective, reversible aldehyde dehydrogenase-2 (ALDH-2) inhibitor with an IC50 of 29 nM. CVT-10216 also has inhibitory effect of ALDH-1 with an IC50 of 1.3 μM. CVT-10216 can reduce excessive alcohol drinking in alcohol-preferring rats and exhibit anxiolytic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0037 Supplier Medchemexpress LLC Supplier No. HY198015MG
Medchemexpress, HY-19801 5mg CVT-10216 CAS:1005334-57-5 CVT-10216 is a highly selective, reversible aldehyde dehydrogenase-2 (ALDH-2) inhibitor with an IC50 of 29 nM. CVT-10216 also has inhibitory effect of ALDH-1 with an IC50 of 1.3 μM. CVT-10216 can reduce excessive alcohol drinking in alcohol-preferring rats and exhibit anxiolytic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.