Learn More
Medchemexpress LLC CVT-12012 | 1018675-35-8 | 99.58% | 434.41 | 10 MG
SDP
Catalog No. 5000344107
Encompass_Preferred
CVT-12012 is a potent and orally bioavailable stearoyl-CoA desaturase (SCD) inhibitor. It is suitable for research purposes and shows high potency in SCD assays.
  • High potency in SCD assays
  • Good oral bioavailability
  • Suitable for research purposes
  • Shows highest potency in both microsomal and HEPG2 SCD assays
  • Oral absorption not significantly affected by Pgp efflux
Catalog No. 50-003-44107 Supplier Medchemexpress LLC Supplier No. HY1103450MG
CVT-12012 is a potent and orally bioavailable stearoyl-CoA desaturase (SCD) inhibitor. It is suitable for research purposes and shows high potency in SCD assays.
  • High potency in SCD assays
  • Good oral bioavailability
  • Suitable for research purposes
  • Shows highest potency in both microsomal and HEPG2 SCD assays
  • Oral absorption not significantly affected by Pgp efflux