Learn More
Medchemexpress LLC CVT-12012 | 1018675-35-8 | 99.58% | 434.41 | 25 MG
SDP
Catalog No. 5000345764
Encompass_Preferred
CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM for rat microsomal and 6.1 nM for human HEPG2.
  • Potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor.
  • Demonstrates good oral bioavailability (78%) in a rat PK study.
  • Oral absorption is not significantly affected by Pgp efflux.
Catalog No. 50-003-45764 Supplier Medchemexpress LLC Supplier No. 25MGHY11034
CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM for rat microsomal and 6.1 nM for human HEPG2.
  • Potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor.
  • Demonstrates good oral bioavailability (78%) in a rat PK study.
  • Oral absorption is not significantly affected by Pgp efflux.