Learn More
Medchemexpress LLC HY-11034 10mg Medchemexpress, CVT-12012 CAS:1018675-35-8 Purity:>98%
SDP
Catalog No. 501968791
Encompass_Preferred
Medchemexpress, HY-11034 10mg CVT-12012 CAS:1018675-35-8 CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM, 6.1 nM for rat microsomal and human HEPG2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8791 Supplier Medchemexpress LLC Supplier No. HY1103410MG
Medchemexpress, HY-11034 10mg CVT-12012 CAS:1018675-35-8 CVT-12012 is a potent and orally bioavailable stearoyl-coA desaturase (SCD) inhibitor, with IC50s of 38 nM, 6.1 nM for rat microsomal and human HEPG2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.