Learn More
Selleck Chemical LLC CVT-313
SDP
Catalog No. 5000002787
Encompass_Preferred
CVT-313 is a potent CDK2 inhibitor with an IC50 of 0 5 microM in vitro It has no effect on other nonrelated ATP-dependent serine/threonine kinases
Catalog No. 50-000-02787 Supplier Selleck Chemical LLC Supplier No. S653710MM/1ML
Add to Cart
Add to Cart
CVT-313 is a potent CDK2 inhibitor with an IC50 of 0 5 microM in vitro It has no effect on other nonrelated ATP-dependent serine/threonine kinases