Learn More
Enzo Life Sciences CVT-313 (5mg). CAS: 199986-75-9
SDP
Supplier:  Enzo Life Sciences ALX270393M005
Encompass_Preferred
Specific inhibitor of CDK2/cyclin A (IC50=500nM) and CDK2/cyclin E (IC50=500nM). Also inhibits CDK1/cyclin B (IC50=4.2µM) and CDK4/cyclin D1 (IC50=215µM). Alternative name: 6-(4-Methoxybenzylamino)-2-[bis(2-hydroxyethylamino)]-9-isopropylpurine. Purity: ≥98% (HPLC). Appearance: White crystalline solid. Solubility: Soluble in DMSO, dimethyl formamide or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8532