Learn More
Apexbio Technology LLC CVT-313,5mg CAS# 199986-75-9
SDP
Catalog No. 502240209
Encompass_Preferred
CVT-313 is a potent inhibitor of cyclin-dependent kinase 2 (CDK2) with IC50 value of 0.5 μM [1].CDK2 is a serine/threonine kinase that is essential for the G1-S phase during cell division. CDK2 is an important target for prevention of aberrant cell proliferation [1].In MRC-5 cells, CVT-313 inhibited Rb hyperphosphorylation in a time-dependant way and the cell cycle was arrested at the G1/S phase. Also, CVT-313 inhibited the growth of mouse, rat and human cells with IC50 values from 1.25 to 20 mM [1]. In human diffuse large B-cell lymphoma (DLBCL) cells, CVT-313 reduced CDK2-mediated phosphorylation of the retinoblastoma gene product (Rb) on T821. Also, CVT-313 reduced the anti-apoptotic factor Myeloid cell leukemia-1 (Mcl-1) and induced apoptosis [2]. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0209 Supplier Apexbio Technology LLC Supplier No. A33365
CVT-313 is a potent inhibitor of cyclin-dependent kinase 2 (CDK2) with IC50 value of 0.5 μM [1].CDK2 is a serine/threonine kinase that is essential for the G1-S phase during cell division. CDK2 is an important target for prevention of aberrant cell proliferation [1].In MRC-5 cells, CVT-313 inhibited Rb hyperphosphorylation in a time-dependant way and the cell cycle was arrested at the G1/S phase. Also, CVT-313 inhibited the growth of mouse, rat and human cells with IC50 values from 1.25 to 20 mM [1]. In human diffuse large B-cell lymphoma (DLBCL) cells, CVT-313 reduced CDK2-mediated phosphorylation of the retinoblastoma gene product (Rb) on T821. Also, CVT-313 reduced the anti-apoptotic factor Myeloid cell leukemia-1 (Mcl-1) and induced apoptosis [2]. Other sizes are also available. Please inqury us for quote.