Learn More
Medchemexpress LLC Cvt-313 (cdk2 inhibitor iii) | 199986-75-9 | C20H28N6O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15339100MG
Encompass_Preferred
CVT-313 (Cdk2 Inhibitor III) is a potent, selective, reversible, and ATP-competitive inhibitor of CDK2 with an IC50 of 0.5 μM. It inhibits CDC5L phosphorylation and exhibits profound effects on cell proliferation. This product is for research use only.
  • Potent, selective, reversible, and ATP-competitive inhibitor of CDK2
  • IC50 of 0.5 μM for CDK2
  • Inhibits CDC5L phosphorylation
  • Exhibits profound effects on cell proliferation at 5-20 μM
  • For research use only
Catalog No. 50-003-72829