Learn More
Medchemexpress LLC CVT-313 (Cdk2 Inhibitor III) | 199986-75-9 | MFCD06411412 | C20H28N6O3 | 1 ML
SDP
Catalog No. 5000373524
Encompass_Preferred
CVT-313 (Cdk2 Inhibitor III) is a potent, selective, reversible, and ATP-competitive inhibitor of CDK2. It has an IC50 of 0.5 μM for CDK2 and inhibits CDC5L phosphorylation. This compound shows significant effects on cell proliferation at concentrations ranging from 5-20 μM, with high selectivity against nonrelated ATP-dependent serine/threonine kinases.
  • Potent and selective CDK2 inhibitor
  • Reversible and ATP-competitive
  • Inhibits CDC5L phosphorylation
  • Active in cell proliferation assays
Catalog No. 50-003-73524 Supplier Medchemexpress LLC Supplier No. HY1533910MM/1ML
CVT-313 (Cdk2 Inhibitor III) is a potent, selective, reversible, and ATP-competitive inhibitor of CDK2. It has an IC50 of 0.5 μM for CDK2 and inhibits CDC5L phosphorylation. This compound shows significant effects on cell proliferation at concentrations ranging from 5-20 μM, with high selectivity against nonrelated ATP-dependent serine/threonine kinases.
  • Potent and selective CDK2 inhibitor
  • Reversible and ATP-competitive
  • Inhibits CDC5L phosphorylation
  • Active in cell proliferation assays