Learn More
Medchemexpress LLC HY-15857 10mg , CW-069 CAS:1594094-64-0 Purity:98%
SDP
Catalog No. 501494071
Encompass_Preferred
Medchemexpress, HY-15857 10mg CW-069 CAS:1594094-64-0 Formula:C23H21IN2O3 IC50: 75 μM (HSET) Purity:98% CW-069 is an allosteric inhibitor of HSET with an IC 50 of 75 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4071 Supplier Medchemexpress LLC Supplier No. HY1585710MG
Medchemexpress, HY-15857 10mg CW-069 CAS:1594094-64-0 Formula:C23H21IN2O3 IC50: 75 μM (HSET) Purity:98% CW-069 is an allosteric inhibitor of HSET with an IC 50 of 75 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.