Learn More
Apexbio Technology LLC CW069, 2mg. Cas: 1594094-64-0 MFCD: MFCD28009372. allosteric inhibitor of HSET, selective
SDP
Catalog No. 501015296
Encompass_Preferred
CW069 is an allosteric, and selective inhibitor of microtubule motor protein HSET with IC50 of 75 uM, significant selectivity over KSP.. MFCD28009372
Catalog No. 50-101-5296 Supplier Apexbio Technology LLC Supplier No. B30622
CW069 is an allosteric, and selective inhibitor of microtubule motor protein HSET with IC50 of 75 uM, significant selectivity over KSP.. MFCD28009372