Learn More
Medchemexpress LLC HY-111746 10mg Medchemexpress, CWHM-1008 CAS: Purity:>98%
SDP
Catalog No. 501968946
Encompass_Preferred
Medchemexpress, HY-111746 10mg CWHM-1008 CAS: CWHM-1008 is a potent and orally active antimalarial agent, with EC50 values of 46 and 21 nM against drug-sensitive Plasmodium falciparum 3D7 and drug-resistant Dd2 strains, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8946 Supplier Medchemexpress LLC Supplier No. HY11174610MG
Medchemexpress, HY-111746 10mg CWHM-1008 CAS: CWHM-1008 is a potent and orally active antimalarial agent, with EC50 values of 46 and 21 nM against drug-sensitive Plasmodium falciparum 3D7 and drug-resistant Dd2 strains, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.