Learn More
Medchemexpress LLC CWHM-1008 | 2362539-97-5 | 99.1% | 405.46 | 1 ML
SDP
Catalog No. 5000439218
Encompass_Preferred
CWHM-1008 is a potent and orally active antimalarial agent. It exhibits EC50 values of 46 nM against agent-sensitive Plasmodium falciparum 3D7 and 21 nM against drug-resistant Dd2 strains. It is for research use only.
  • Potent and orally active antimalarial agent.
  • Effective against both agent-sensitive Plasmodium falciparum 3D7 and drug-resistant Dd2 strains.
Catalog No. 50-004-39218 Supplier Medchemexpress LLC Supplier No. HY11174610MM/1ML
CWHM-1008 is a potent and orally active antimalarial agent. It exhibits EC50 values of 46 nM against agent-sensitive Plasmodium falciparum 3D7 and 21 nM against drug-resistant Dd2 strains. It is for research use only.
  • Potent and orally active antimalarial agent.
  • Effective against both agent-sensitive Plasmodium falciparum 3D7 and drug-resistant Dd2 strains.