Learn More
Medchemexpress LLC CWHM-1008 | 2362539-97-5 | 99.1% | 405.46 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111746
Encompass_Preferred
CWHM-1008 is a potent and orally active antimalarial agent. It exhibits EC50 values of 46 nM against agent-sensitive Plasmodium falciparum 3D7 and 21 nM against drug-resistant Dd2 strains.
  • Potent antimalarial agent
  • Orally active
  • Effective against both agent-sensitive and drug-resistant strains of Plasmodium falciparum
Catalog No. 50-004-39916