Learn More
Medchemexpress LLC HY-18644 100mg , CWHM-12 CAS:1564286-55-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY18644100MG
Encompass_Preferred
Medchemexpress, HY-18644 100mg CWHM-12 CAS:1564286-55-0 Formula:C26H32BrN5O6 IC50: 0.2 nM (αvβ8), 0.8 nM (αvβ3), 1.5 nM (αvβ6), 1.8 nM (αvβ1), 61 nM (αvβ5) Purity:98% CWHM-12 is a potent inhibitor of αV integrins with IC 50 s of 0.2, 0.8, 1.5, and 1.8 nM for αvβ8 , αvβ3 , αvβ6 , and αvβ1 . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3040