Learn More
TARGETMOL CHEMICALS INC CWHM-1552 5MG
SDP
Catalog No. 502581621
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CWHM-1552 is an effective Plasmodium falciparum inhibitor. For the 3D7 and Dd2 strains the IC50s are 51 nM and 53 nM. purity: 100%
Catalog No. 50-258-1621 Supplier TARGETMOL CHEMICALS INC Supplier No. T109035MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CWHM-1552 is an effective Plasmodium falciparum inhibitor. For the 3D7 and Dd2 strains the IC50s are 51 nM and 53 nM. purity: 100%