Learn More
Medchemexpress LLC CWI1-2 hydrochloride | 2408590-37-2 | C22H18Cl4N6O3 | 25 MG
SDP
Catalog No. 5000375494
Encompass_Preferred
CWI1-2 hydrochloride is an IGF2BP2 inhibitor that binds IGF2BP2 and inhibits its interaction with m6A-modified target transcripts. It induces apoptosis and differentiation, and demonstrates promising anti-leukemic effects. It is intended for research use only.
  • Binds IGF2BP2
  • Inhibits interaction with m6A-modified target transcripts
  • Induces apoptosis
  • Induces differentiation
  • Demonstrates anti-leukemic effects
  • For research use only
Catalog No. 50-003-75494 Supplier Medchemexpress LLC Supplier No. 25MGHY153274A
CWI1-2 hydrochloride is an IGF2BP2 inhibitor that binds IGF2BP2 and inhibits its interaction with m6A-modified target transcripts. It induces apoptosis and differentiation, and demonstrates promising anti-leukemic effects. It is intended for research use only.
  • Binds IGF2BP2
  • Inhibits interaction with m6A-modified target transcripts
  • Induces apoptosis
  • Induces differentiation
  • Demonstrates anti-leukemic effects
  • For research use only