Learn More
Medchemexpress LLC CWI1-2 hydrochloride | 2408590-37-2 | C22H18Cl4N6O3 | 5 MG
SDP
Catalog No. 5000372545
Encompass_Preferred
CWI1-2 hydrochloride is an IGF2BP2 inhibitor that binds IGF2BP2 and inhibits its interaction with m6A-modified target transcripts, induces apoptosis and differentiation, and shows promising anti-leukemic effects.
  • IGF2BP2 inhibitor.
  • Inhibits IGF2BP2 interaction with m6A-modified target transcripts.
  • Induces apoptosis and differentiation.
  • Demonstrates anti-leukemic effects.
  • Effective in reducing Gln uptake and impairing mitochondrial function in AML cells.
  • Inhibits colony-forming ability of leukemic mouse blasts.
  • Impairs self-renewal of LSC/LIC.
  • Delays leukemia onset and prolongs survival in vivo.
Catalog No. 50-003-72545 Supplier Medchemexpress LLC Supplier No. HY153274A5MG
CWI1-2 hydrochloride is an IGF2BP2 inhibitor that binds IGF2BP2 and inhibits its interaction with m6A-modified target transcripts, induces apoptosis and differentiation, and shows promising anti-leukemic effects.
  • IGF2BP2 inhibitor.
  • Inhibits IGF2BP2 interaction with m6A-modified target transcripts.
  • Induces apoptosis and differentiation.
  • Demonstrates anti-leukemic effects.
  • Effective in reducing Gln uptake and impairing mitochondrial function in AML cells.
  • Inhibits colony-forming ability of leukemic mouse blasts.
  • Impairs self-renewal of LSC/LIC.
  • Delays leukemia onset and prolongs survival in vivo.