Learn More
MSC Tiger-Vac Cleanroom Vacuum, Electric, ULPA Filter, 7.5 gal, 1 ea
Catalog No. 502819227
Encompass
This electric ULPA filtered vacuum cleaner is designed for cleanroom, electronics, and ESD-safe environments, capable of both dry and wet cleaning. It features a durable 7.5 gal stainless steel tank and operates efficiently for various industrial applications.
  • ULPA filtration for critical environments
  • Capacity of 7.5 gal stainless steel tank
  • Suitable for dry and wet vacuuming
  • ESD-safe for sensitive electronics
  • Includes a comprehensive tool kit
  • Meets CSA and UL standards
Catalog No. 50-281-9227 Supplier MSC Supplier No. 28989135
Add to Cart
Add to Cart
Request a Quote
This electric ULPA filtered vacuum cleaner is designed for cleanroom, electronics, and ESD-safe environments, capable of both dry and wet cleaning. It features a durable 7.5 gal stainless steel tank and operates efficiently for various industrial applications.
  • ULPA filtration for critical environments
  • Capacity of 7.5 gal stainless steel tank
  • Suitable for dry and wet vacuuming
  • ESD-safe for sensitive electronics
  • Includes a comprehensive tool kit
  • Meets CSA and UL standards