Learn More
Medchemexpress LLC HY-50855 100mg Medchemexpress, CX-4945 Silmitasertib CAS:1009820-21-6 Purity:98%
SDP
Catalog No. 501639023
Encompass_Preferred
Medchemexpress,HY-50855 100mg CX-4945 Silmitasertib CAS:1009820-21-6 Formula:C19H12ClN3O2 CX-4945 is a potent, selective and oral casein kinase 2 (CK2) inhibitor with a Ki of 0.38 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9023 Supplier Medchemexpress LLC Supplier No. HY50855100MG
Medchemexpress,HY-50855 100mg CX-4945 Silmitasertib CAS:1009820-21-6 Formula:C19H12ClN3O2 CX-4945 is a potent, selective and oral casein kinase 2 (CK2) inhibitor with a Ki of 0.38 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.