Learn More
Apexbio Technology LLC CX-4945 (Silmitasertib) 1009820-21-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062226
Encompass_Preferred
CX-4945 (Silmitasertib CAS 1009820-21-6) is an orally bioavailable inhibitor targeting casein kinase 2 (CK2) via ATP-competitive binding with an IC50 of approximately 1 nM It suppresses CK2-mediated signaling pathways notably by reducing phosphorylation of Akt at Ser129 independently of PTEN activation thus impacting PI3K/Akt pathway regulation In vitro studies report that CX-4945 treatment leads to increased total expression and decreased phosphorylation of cell cycle regulators p21 and p27 causing cell cycle arrest (G2/M in BT-474 breast carcinoma G1 in BxPC-3 pancreatic carcinoma) In vivo it displays antitumor activity in BxPC-3 mouse xenograft models highlighting its utility for oncology research
Catalog No. 50-000-62226 Supplier Apexbio Technology LLC Supplier No. A83305.1
CX-4945 (Silmitasertib CAS 1009820-21-6) is an orally bioavailable inhibitor targeting casein kinase 2 (CK2) via ATP-competitive binding with an IC50 of approximately 1 nM It suppresses CK2-mediated signaling pathways notably by reducing phosphorylation of Akt at Ser129 independently of PTEN activation thus impacting PI3K/Akt pathway regulation In vitro studies report that CX-4945 treatment leads to increased total expression and decreased phosphorylation of cell cycle regulators p21 and p27 causing cell cycle arrest (G2/M in BT-474 breast carcinoma G1 in BxPC-3 pancreatic carcinoma) In vivo it displays antitumor activity in BxPC-3 mouse xenograft models highlighting its utility for oncology research