Learn More
Selleck Chemical LLC Silmitasertib (CX-4945) 5mg 1009820-21-6
SDP
Catalog No. 506959
Encompass_Preferred
Silmitasertib (CX-4945) is a potent and selective inhibitor of CK2 (casein kinase 2) with IC50 of 1 nM in a cell-free assay, less potent to Flt3, Pim1 and CDK1 (inactive in cell-based assay). Silmitasertib induces autophagy and promotes apoptosis. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-695-9 Supplier Selleck Chemical LLC Supplier No. S22485MG
Add to Cart
Add to Cart
Silmitasertib (CX-4945) is a potent and selective inhibitor of CK2 (casein kinase 2) with IC50 of 1 nM in a cell-free assay, less potent to Flt3, Pim1 and CDK1 (inactive in cell-based assay). Silmitasertib induces autophagy and promotes apoptosis. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.