Learn More
Apexbio Technology LLC CX-4945 sodium salt 1309357-15-0 50mg
SDP
Catalog No. 5000091139
Encompass_Preferred
CX-4945 sodium salt (Silmitasertib sodium salt CAS 1309357-15-0) is a potent and selective inhibitor of casein kinase 2 (CK2) effectively targeting both the CK2 and CK2 catalytic subunits By inhibiting CK2 activity CX-4945 disrupts signaling pathways involved in cell cycle progression transcriptional regulation and cell survival The compound exhibits oral bioavailability and is widely utilized in research focused on tumor biology mechanistic studies of signaling cascades and the functional characterization of protein kinases
Catalog No. 50-000-91139 Supplier Apexbio Technology LLC Supplier No. B490350
CX-4945 sodium salt (Silmitasertib sodium salt CAS 1309357-15-0) is a potent and selective inhibitor of casein kinase 2 (CK2) effectively targeting both the CK2 and CK2 catalytic subunits By inhibiting CK2 activity CX-4945 disrupts signaling pathways involved in cell cycle progression transcriptional regulation and cell survival The compound exhibits oral bioavailability and is widely utilized in research focused on tumor biology mechanistic studies of signaling cascades and the functional characterization of protein kinases