Learn More
Apexbio Technology LLC CX-4945 sodium salt 1309357-15-0 5mg
SDP
Catalog No. 5000087946
Encompass_Preferred
CX-4945 sodium salt (Silmitasertib sodium salt CAS 1309357-15-0) is a potent and selective inhibitor of casein kinase 2 (CK2) effectively targeting both the CK2 and CK2 catalytic subunits By inhibiting CK2 activity CX-4945 disrupts signaling pathways involved in cell cycle progression transcriptional regulation and cell survival The compound exhibits oral bioavailability and is widely utilized in research focused on tumor biology mechanistic studies of signaling cascades and the functional characterization of protein kinases
Catalog No. 50-000-87946 Supplier Apexbio Technology LLC Supplier No. B49035
CX-4945 sodium salt (Silmitasertib sodium salt CAS 1309357-15-0) is a potent and selective inhibitor of casein kinase 2 (CK2) effectively targeting both the CK2 and CK2 catalytic subunits By inhibiting CK2 activity CX-4945 disrupts signaling pathways involved in cell cycle progression transcriptional regulation and cell survival The compound exhibits oral bioavailability and is widely utilized in research focused on tumor biology mechanistic studies of signaling cascades and the functional characterization of protein kinases