Learn More
Medchemexpress LLC CX-5011 | 1333382-30-1 | 99.5% | C20H11N4NaO2 | 5 MG
SDP
Catalog No. 5000359289
Encompass_Preferred
CX-5011 is a CK2 inhibitor that induces Rac1 activation, apoptosis, and cancer cell death.
  • CK2 inhibitor
  • Induces Rac1 activation
  • Induces apoptosis
  • Leads to cancer cell death
Catalog No. 50-003-59289 Supplier Medchemexpress LLC Supplier No. HY50733A5MG
CX-5011 is a CK2 inhibitor that induces Rac1 activation, apoptosis, and cancer cell death.
  • CK2 inhibitor
  • Induces Rac1 activation
  • Induces apoptosis
  • Leads to cancer cell death