Learn More
Medchemexpress LLC HY-13323 5mg Medchemexpress, CX-5461 CAS:1138549-36-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY133235MG
Encompass_Preferred
Medchemexpress, HY-13323 5mg CX-5461 CAS:1138549-36-6 CX-5461 is a potent and oral rRNA synthesis inhibitor. It inhibits RNA polymerase I-driven transcription of rRNA with IC50s of 142, 113, and 54 nM in HCT-116, A375, and MIA PaCa-2 cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9852