Learn More
Medchemexpress LLC HY-13323A 5mg Medchemexpress, CX-5461 dihydrochloride CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13323A5MG
Encompass_Preferred
Medchemexpress, HY-13323A 5mg CX-5461 dihydrochloride CAS: CX-5461 dihydrochloride is a potent and orally bioavailable inhibitor of Pol I-mediated rRNA synthesis , with IC 50 s of 142 nM in HCT-116, 113 nM in A375, and 54 nM in MIA PaCa-2 cells, and shows little or no effect on Pol II ( IC 50 ≥25 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2663