Learn More
TARGETMOL CHEMICALS INC CX-6258 HYDROCHLORIDE 10MG
SDP
Catalog No. 502579517
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CX-6258 hydrochloride (Pim-Kinase Inhibitor X) is an effective orally efficacious Pim1/2/3 kinase inhibitor (IC50 5/25/16 nM). purity: 98%
Catalog No. 50-257-9517 Supplier TARGETMOL CHEMICALS INC Supplier No. T614810MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CX-6258 hydrochloride (Pim-Kinase Inhibitor X) is an effective orally efficacious Pim1/2/3 kinase inhibitor (IC50 5/25/16 nM). purity: 98%