Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CX-6258 (hydrochloride) | 1353859-00-3 | 99.0% | 498.40 | 5 MG
SDP

Catalog No. 5000387446
Encompass_Preferred

CX-6258 hydrochloride is a potent and kinase-selective pan-Pim kinases inhibitor, with IC50s of 5 nM, 25 nM, and 16 nM for Pim-1, Pim-2, and Pim-3, respectively.

  • Purity of 99.03%.
  • Solid appearance.
  • Orange to red color.
  • Soluble in DMSO (10 mg/mL, requires ultrasonic).
  • Soluble in H2O (1 mg/mL, requires ultrasonic, warming, and heating to 60°C).
  • In vivo solubility of ≥ 2.5 mg/mL in a specific solution.

Catalog No. 50-003-87446 Supplier Medchemexpress LLC Supplier No. HY18095B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CX-6258 hydrochloride is a potent and kinase-selective pan-Pim kinases inhibitor, with IC50s of 5 nM, 25 nM, and 16 nM for Pim-1, Pim-2, and Pim-3, respectively.

  • Purity of 99.03%.
  • Solid appearance.
  • Orange to red color.
  • Soluble in DMSO (10 mg/mL, requires ultrasonic).
  • Soluble in H2O (1 mg/mL, requires ultrasonic, warming, and heating to 60°C).
  • In vivo solubility of ≥ 2.5 mg/mL in a specific solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.