Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CX-6258 hydrochloride | 1353859-00-3 | MFCD28142834 | 99.0% | 498.40 | C26H25Cl2N3O3 | 10 MG
SDP

Catalog No. 5000297970
Encompass_Preferred

CX-6258 hydrochloride is the hydrochloride salt of a potent, kinase-selective pan-Pim inhibitor used for research applications. It exhibits low-nanomolar biochemical potency against Pim family kinases and is supplied with solubility and storage information to support in vitro and in vivo studies.

  • Inhibits Pim-1, Pim-2, and Pim-3 with IC50s of 5 nM, 25 nM, and 16 nM, respectively.
  • High purity (~99%) suitable for biochemical and cell-based assays.
  • Available as a solid and as a 10 mM solution in DMSO for flexible use.
  • Soluble in DMSO (10 mg/mL) and in water with heating (1 mg/mL).
  • Storage recommendations provided for solid and solution to preserve stability.
  • Orange to red solid facilitating visual inspection of material.

Catalog No. 50-002-97970 Supplier Medchemexpress LLC Supplier No. HY18095B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CX-6258 hydrochloride is the hydrochloride salt of a potent, kinase-selective pan-Pim inhibitor used for research applications. It exhibits low-nanomolar biochemical potency against Pim family kinases and is supplied with solubility and storage information to support in vitro and in vivo studies.

  • Inhibits Pim-1, Pim-2, and Pim-3 with IC50s of 5 nM, 25 nM, and 16 nM, respectively.
  • High purity (~99%) suitable for biochemical and cell-based assays.
  • Available as a solid and as a 10 mM solution in DMSO for flexible use.
  • Soluble in DMSO (10 mg/mL) and in water with heating (1 mg/mL).
  • Storage recommendations provided for solid and solution to preserve stability.
  • Orange to red solid facilitating visual inspection of material.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.