Learn More
Medchemexpress LLC CX-6258 hydrochloride hydrate | 1353858-99-7 | 99.2% | 10 MG
SDP
Catalog No. 5000294692
Encompass_Preferred
A research-grade hydrochloride hydrate salt of a potent pan-Pim kinase inhibitor intended for laboratory research use. It selectively inhibits Pim-1, Pim-2, and Pim-3 with reported IC50 values of 5 nM, 25 nM, and 16 nM, respectively. Characterization data and quality documents (SDS, COA, LCMS) are provided by the supplier.
Catalog No. 50-002-94692 Supplier Medchemexpress LLC Supplier No. HY18095A10MG
A research-grade hydrochloride hydrate salt of a potent pan-Pim kinase inhibitor intended for laboratory research use. It selectively inhibits Pim-1, Pim-2, and Pim-3 with reported IC50 values of 5 nM, 25 nM, and 16 nM, respectively. Characterization data and quality documents (SDS, COA, LCMS) are provided by the supplier.