Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CX-6258 hydrochloride hydrate | 1353858-99-7 | 99.2% | 516.42 | 25 MG
SDP

Catalog No. 5000384800
Encompass_Preferred

CX-6258 hydrochloride hydrate is a potent and selective pan-Pim kinases inhibitor. It effectively inhibits Pim-1, Pim-2, and Pim-3, making it suitable for research into cancer and metabolic diseases.

  • Potent and kinase selective pan-Pim kinases inhibitor
  • Inhibits Pim-1 (IC50: 5 nM), Pim-2 (IC50: 25 nM), and Pim-3 (IC50: 16 nM)
  • Causes dose-dependent inhibition of phosphorylation of pro-survival proteins, Bad and 4E-BP1
  • Diminishes steady-state levels of ectopic NKX3.1 in PC3 cells
  • Exhibits robust in vivo efficacy in Pim kinases driven tumor models
  • Achieves 45% tumor growth inhibition at 50 mg/kg and 75% at 100 mg/kg in vivo

Catalog No. 50-003-84800 Supplier Medchemexpress LLC Supplier No. 25MGHY18095A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CX-6258 hydrochloride hydrate is a potent and selective pan-Pim kinases inhibitor. It effectively inhibits Pim-1, Pim-2, and Pim-3, making it suitable for research into cancer and metabolic diseases.

  • Potent and kinase selective pan-Pim kinases inhibitor
  • Inhibits Pim-1 (IC50: 5 nM), Pim-2 (IC50: 25 nM), and Pim-3 (IC50: 16 nM)
  • Causes dose-dependent inhibition of phosphorylation of pro-survival proteins, Bad and 4E-BP1
  • Diminishes steady-state levels of ectopic NKX3.1 in PC3 cells
  • Exhibits robust in vivo efficacy in Pim kinases driven tumor models
  • Achieves 45% tumor growth inhibition at 50 mg/kg and 75% at 100 mg/kg in vivo

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.