Learn More
Medchemexpress LLC CX 717 | 867276-98-0 | 99.9% | 233.22 | 50 MG
SDP
Catalog No. 5000350446
Encompass_Preferred
CX 717 is identified as a positive allosteric modulator of the AMPA receptor, with an antidepressant-like effect.
  • Positive allosteric modulator of AMPA receptor.
  • Exhibits antidepressant-like effects.
  • Can be used for the research of adult attention deficit hyperactivity disorder (ADHD).
  • In vivo studies showed that CX 717 (20 mg/kg) has a rapid (30 min) but short-lasting (up to 24 h) antidepressant-like effect in the forced swim test.
  • Produces a rapid (up to 1 h) increase of brain-derived neurotrophic factor (BDNF) and a more sustained (up to 6 h) increase of p11.
  • Intra-cortical infusion of CX 717 (85 mM) increases the efflux of noradrenaline, dopamine, and serotonin, but not glutamate.
Catalog No. 50-003-50446 Supplier Medchemexpress LLC Supplier No. HY13989750MG
CX 717 is identified as a positive allosteric modulator of the AMPA receptor, with an antidepressant-like effect.
  • Positive allosteric modulator of AMPA receptor.
  • Exhibits antidepressant-like effects.
  • Can be used for the research of adult attention deficit hyperactivity disorder (ADHD).
  • In vivo studies showed that CX 717 (20 mg/kg) has a rapid (30 min) but short-lasting (up to 24 h) antidepressant-like effect in the forced swim test.
  • Produces a rapid (up to 1 h) increase of brain-derived neurotrophic factor (BDNF) and a more sustained (up to 6 h) increase of p11.
  • Intra-cortical infusion of CX 717 (85 mM) increases the efflux of noradrenaline, dopamine, and serotonin, but not glutamate.