Learn More
Mitegen CX100 Dryshipper By IC Biomedical With Upright Shipping Case Bundle, Ideal For -190°C Cryogenic Transport, IATA Compliant
SDP
Catalog No. NC2159060
Encompass
This bundle features the CX100 Cryo Express Dry Shipper by IC Biomedical and a hard plastic Upright Shipping Case, offering a comprehensive solution for safe cryogenic material transport. The CX100 Dry Shipper is engineered to safely move various materials at approximately -190°C. Its unique adsorbent material prevents liquid nitrogen spills even if tipped, ensuring reliable transit and IATA compliance for non-infectious biological samples. The included Upright Shipping Case provides next-generation protection. Made of high-impact-resistant material with a hard plastic shell and padded lining, it safeguards contents from physical damage. Its domed top prevents stacking, and a wide circular base encourages upright orientation during handling. The CX100 offers an 18-day static holding time and 4.1 liters LN2 adsorbed capacity.
This item is not returnable. View return policy
Catalog No. NC2159060 Supplier Mitegen Supplier No. TWCXS2
This bundle features the CX100 Cryo Express Dry Shipper by IC Biomedical and a hard plastic Upright Shipping Case, offering a comprehensive solution for safe cryogenic material transport. The CX100 Dry Shipper is engineered to safely move various materials at approximately -190°C. Its unique adsorbent material prevents liquid nitrogen spills even if tipped, ensuring reliable transit and IATA compliance for non-infectious biological samples. The included Upright Shipping Case provides next-generation protection. Made of high-impact-resistant material with a hard plastic shell and padded lining, it safeguards contents from physical damage. Its domed top prevents stacking, and a wide circular base encourages upright orientation during handling. The CX100 offers an 18-day static holding time and 4.1 liters LN2 adsorbed capacity.