Learn More
Aviva Systems Biology Corp CX36 Antibody - middle region (ARP36685_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36685_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CX36. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CX36. Alternate names: CX36, GJA9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56249