Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CX3CL1 Protein, His Tag, ultra sensitivity (primary amine labeling)
Catalog No. 501957543
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CX3CL1, His Tag, primary amine labeling (CX1-H8221) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 341 (Accession # AAH01163). It is the biotinylated form of Human CX3CL1, His Tag (Cat. No.CX1-H5221).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.5 kDa. The protein migrates as 50-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7543 Supplier ACROBiosystems Supplier No. CX1H822125UG
Recombinant Protein;25UG;Biotinylated Human CX3CL1, His Tag, primary amine labeling (CX1-H8221) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 341 (Accession # AAH01163). It is the biotinylated form of Human CX3CL1, His Tag (Cat. No.CX1-H5221).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.5 kDa. The protein migrates as 50-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.