Learn More
Medchemexpress LLC CX516 | 154235-83-3 | 99.94% | 241.29 | 100 MG
SDP
Catalog No. 5000344702
Encompass_Preferred
CX516 (BDP 12) is an ampakine that functions as an AMPA receptor positive allosteric modulator. It is used in the research of Alzheimer's disease, schizophrenia, and mild cognitive impairment (MCI).
  • Acts as an AMPA receptor positive allosteric modulator.
  • Used in research for Alzheimer's disease, schizophrenia, and mild cognitive impairment (MCI).
Catalog No. 50-003-44702 Supplier Medchemexpress LLC Supplier No. HY10933200MG
CX516 (BDP 12) is an ampakine that functions as an AMPA receptor positive allosteric modulator. It is used in the research of Alzheimer's disease, schizophrenia, and mild cognitive impairment (MCI).
  • Acts as an AMPA receptor positive allosteric modulator.
  • Used in research for Alzheimer's disease, schizophrenia, and mild cognitive impairment (MCI).