Learn More
Sino Biological CXADR / CAR Antibody (PE), Rabbit MAb 25Tests
Supplier:  Sino Biological 10799R271P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CXADR / CAR (rh CXADR / CAR; Catalog#10799-H08H; NP_001329.1; Met1-Gly237) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-224-8083