Learn More
Cayman Chemical CXADR RabbIt MoncLNal ntIb 1mL
SDP
Catalog No. 502916592
Encompass_Preferred
Immunogen: Recombinant human CXADR8 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) CXADR • Applications: ELISA, FC, ICC, IF
Catalog No. 50-291-6592 Supplier Cayman Chemical Supplier No. 380841 ML
Immunogen: Recombinant human CXADR8 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) CXADR • Applications: ELISA, FC, ICC, IF