Learn More
ACROBiosystems Human CXADR Protein, His Tag, HEK293, 100 μg
Catalog No. 501657797
Encompass_Preferred
Human CXADR, also known as CAR, is a type I transmembrane glycoprotein belonging to the CTX family of the Ig superfamily. It serves as a receptor for group B coxsackie viruses and subgroup C adenoviruses and functions as a cell adhesion molecule. This protein is expressed in HEK293 cells and contains a C-terminal polyhistidine tag.
  • Greater than 95% purity as determined by SDS-PAGE.
  • Calculated molecular weight of 24.9 kDa, migrating as 32-35 kDa under reducing conditions due to glycosylation.
  • Lyophilized from a 0.22 μm filtered solution in PBS, pH 7.4 with trehalose as a protectant.
  • Endotoxin level is less than 1.0 EU per μg by the LAL or rFC method.
  • Stable for 12 months at -20°C to -70°C in lyophilized state.
Catalog No. 50-165-7797 Supplier ACROBiosystems Supplier No. CXRH5222100UG
Human CXADR, also known as CAR, is a type I transmembrane glycoprotein belonging to the CTX family of the Ig superfamily. It serves as a receptor for group B coxsackie viruses and subgroup C adenoviruses and functions as a cell adhesion molecule. This protein is expressed in HEK293 cells and contains a C-terminal polyhistidine tag.
  • Greater than 95% purity as determined by SDS-PAGE.
  • Calculated molecular weight of 24.9 kDa, migrating as 32-35 kDa under reducing conditions due to glycosylation.
  • Lyophilized from a 0.22 μm filtered solution in PBS, pH 7.4 with trehalose as a protectant.
  • Endotoxin level is less than 1.0 EU per μg by the LAL or rFC method.
  • Stable for 12 months at -20°C to -70°C in lyophilized state.