Learn More
ACROBiosystems Recombinant Protein;Human CXADR / CAR Protein;HEK293;1MG;CXR,CXADR,CAR,CAR4/6,HCAR
Supplier:  ACROBiosystems CXRH52221MG
Encompass_Preferred
Recombinant Protein;Human CXADR / CAR Protein;1MG;Human CXADR, His Tag (CXR-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 237 (Accession # NP_001329.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.9 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7798