Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier: enQuire Bioreagents QP521610UG
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: E. coli. Species Reactivity: Mouse. BioActivity: Mouse CXCL12 / SDF-1 Isoform beta induces primary human T cell chemotaxis (significanty different from control at Mouse CXCL12 / SDF-1 Isoform beta concentrations greater than 5-50 ng/ml). AA Sequence: KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KM. Buffer: Sterile filtered (0.22 um) solution containing 0.1% TFA. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: CXCL12. Uniprot: P40224-2.