Learn More
Aviva Systems Biology Corp CXCL14 Antibody - middle region (AVARP07023_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP07023_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CXCL14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CXCL14. Alternate names: KEC, KS1, BMAC, BRAK, NJAC, MIP2G, MIP-2g, SCYB14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Rat
Catalog No. 50-014-53774