Learn More
Description
Specifications
Specifications
| Description | chemokine (C-X-C motif) ligand 14 |
| Cross Reactivity | Bovine |
| Target Species | Bovine |
| For Use With (Application) | Quantitative |
| Gene ID (Entrez) | 511771 |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Storage Requirements | Store at 4°C for six months. For long term storage store at -20°C. Avoid repeated freezing and thawing. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.