Learn More
Invitrogen™ Mouse CXCL16 ELISA Kit
Description
The Mouse CXCL16 ELISA quantitates Ms CXCL16 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms CXCL16. Principle of the method The Mouse CXCL16 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | Q8BSU2 |
| Assay Range | 1.3-80 pg/mL |
| Assay Sensitivity | 1.3 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Protein Family | Chemokines & Receptors |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Mouse |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.