Learn More
Medchemexpress LLC CXCL15 Mouse HEK29 50ug
SDP
Catalog No. 5000182360
Encompass_Preferred
CXCL15 also known as Lungkine or WECHE is a member of the ELR motif-containing CXC chemokines CXCL15 has neutrophil chemotactic activity CXCL15 is high expression in the lungs of mice[1][2] CXCL15 Protein Mouse (HEK293 His) is produced in HEK293 cells with six C-Terminal His-tags It consists of 142 amino acids (Q26-A167)
Catalog No. 50-001-82360 Supplier Medchemexpress LLC Supplier No. HYP7268250UG
CXCL15 also known as Lungkine or WECHE is a member of the ELR motif-containing CXC chemokines CXCL15 has neutrophil chemotactic activity CXCL15 is high expression in the lungs of mice[1][2] CXCL15 Protein Mouse (HEK293 His) is produced in HEK293 cells with six C-Terminal His-tags It consists of 142 amino acids (Q26-A167)