Learn More
Medchemexpress LLC HY-P7152 10ug Medchemexpress, CXCL3 (CHO-expressed), Rat CAS: Purity:>98%
SDP
Catalog No. 502083117
Encompass_Preferred
Medchemexpress, HY-P7152 10ug CXCL3 (CHO-expressed), Rat CAS: Recombinant Rat CXCL3 (CHO-expressed) acts as a chemoattractant for neutrophils and is involved in the inflammatory response. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3117 Supplier Medchemexpress LLC Supplier No. HYP715210UG
Medchemexpress, HY-P7152 10ug CXCL3 (CHO-expressed), Rat CAS: Recombinant Rat CXCL3 (CHO-expressed) acts as a chemoattractant for neutrophils and is involved in the inflammatory response. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.